Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

65420

Sigma-Aldrich

4-Methoxycinnamic acid

≥98.0% (GC)

Synonym(s):

trans-3-(4-Methoxyphenyl)acrylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O3
CAS Number:
Molecular Weight:
178.18
Beilstein/REAXYS Number:
2208397
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.0% (GC)

mp

173.5 °C (lit.)

SMILES string

COc1ccc(\C=C\C(O)=O)cc1

InChI

1S/C10H10O3/c1-13-9-5-2-8(3-6-9)4-7-10(11)12/h2-7H,1H3,(H,11,12)/b7-4+

InChI key

AFDXODALSZRGIH-QPJJXVBHSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H V Meyers et al.
Carbohydrate research, 197, 15-32 (1990-03-25)
This study completes the spectroscopic basis for a novel oligosaccharide microanalytical method, wherein a derivatization sequence provides monosaccharide subunits bearing two types of exciton-coupling chromophore groups ("bichromophoric") for circular dichroic spectroscopy, namely 4-bromobenzoate (lambda max 245 nm) and 4-methoxycinnamate (lambda
Yoshio Nakagawa et al.
Biochemical pharmacology, 66(1), 63-73 (2003-06-24)
The metabolism and action of trans-anethole (anethole) and the estrogen-like activity of the compound and its metabolites were studied in freshly isolated rat hepatocytes and cultured MCF-7 human breast cancer cells, respectively. The incubation of hepatocytes with anethole (0.25-2.0mM) caused
Sirichai Adisakwattana et al.
International journal of molecular sciences, 13(2), 1778-1789 (2012-03-13)
Cinnamic acid and its derivatives have shown a variety of pharmacologic properties. However, little is known about the antiglycation properties of cinnamic acid and its derivatives. The present study sought to characterize the protein glycation inhibitory activity of cinnamic acid
H M Chawla et al.
Journal of photochemistry and photobiology. B, Biology, 105(1), 25-33 (2011-08-02)
A series of novel calix[4]arene enones (5-7) and cinnamates (12-14) have been synthesized and evaluated for ensuring protection from ultraviolet radiation (UVR). Spectroscopic analyses has revealed that compound 6 absorbs ultraviolet radiations between 280 and 350 nm with an absorption
Victor S Sobolev et al.
Journal of agricultural and food chemistry, 54(10), 3505-3511 (2006-05-17)
The peanut plant (Arachis hypogaea) is known to produce stilbene phytoalexins as a defensive response to fungal invasion; however, the distribution of phytoalexins among different organs of the peanut plant at early stages of growth under axenic conditions has not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service