Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

648043

Sigma-Aldrich

PAMAM dendrimer, cystamine core, generation 4, solution

10 wt. % in methanol

Synonym(s):

PAMAM dendrimer, cystamine core, generation 4.0 solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2CH2CH2SSCH2CH2NH2]:(G=4);dendri PAMAM(NH2)64
Molecular Weight:
14306.35
MDL number:
UNSPSC Code:
12162002

description

amino surface groups

form

solution

mol wt

generation 4

feature

core type cystamine core

concentration

10 wt. % in methanol

no. Surface Groups

64

bp

64.7 °C

density

0.860 g/mL at 25 °C

storage temp.

−20°C

Packaging

unit weight includes solvent

Legal Information

Mfg. by Dendritic NanoTechnologies Inc. Licensed for research use only

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huang, B. H., Tomalia, D. A.
Journal of Luminescence, 111, 215-223 (2005)
DeMattei, C. R., Huang, B. H., Tomalia, D. A.
Nano Letters, 4, 771-777 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service