Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

643602

Sigma-Aldrich

Piperidine on Rasta Resin

50-100 mesh, extent of labeling: 3.0-5.0 mmol/g loading, 2 % cross-linked

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
PubChem Substance ID:

crosslinking

2 % cross-linked

extent of labeling

3.0-5.0 mmol/g loading

particle size

50-100 mesh

storage temp.

2-8°C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service