Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

643181

Sigma-Aldrich

Palladium

extent of labeling: 5 wt. % loading (dry basis), matrix activated carbon, wet support, Degussa type E105CA/W

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd
CAS Number:
Molecular Weight:
106.42
MDL number:
UNSPSC Code:
12141733
PubChem Substance ID:

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

extent of labeling

5 wt. % loading (dry basis)

resistivity

9.96 μΩ-cm, 20°C

matrix

activated carbon, wet support

bp

2970 °C (lit.)

mp

1554 °C (lit.)

density

12.02 g/cm3 (lit.)

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for hydrogenation of alkenes, alkynes, ketones, nitriles,imines, azides, nitro groups, benzenoid and heterocyclic aromatics;used for hydrogenolysis of cyclopropanes, benzyl derivatives,epoxides, hydrazines, and halides; used to dehydrogenate aromatics and deformylate aldehydes.
Heterogeneous catalyst for carbon-carbon coupling reactions (Heck and Suzuki)
It can be used along with ligandless palladium on activated carbon in the preparation of 1-biphenyl-4-yl-ethanone via Suzuki-Miyaura cross-coupling of phenylboronic acid with 4-chloro-acetophenone in water.[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suzuki-Miyaura cross-coupling of aryl chlorides in water using ligandless palladium on activated carbon.
Lysen M and Kohler K.
Synlett, 2005(11), 1671-1674 (2005)
Meifang Zheng et al.
Organic letters, 15(8), 1838-1841 (2013-04-02)
Palladium-catalyzed oxidative difunctionalization of enol ethers with 1,3-dicarbonyl compounds to construct trisubstituted furans in one step under mild conditions is described. The reaction is thought to proceed through a C-C bond formation along with a C-O bond closing the ring
Xiaohu Xia et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(17), 6669-6673 (2013-04-10)
Controlling the shape or morphology of metal nanocrystals is central to the realization of their many applications in catalysis, plasmonics, and electronics. In one of the approaches, the metal nanocrystals are grown from seeds of certain crystallinity through the addition
V Volarevic et al.
Journal of B.U.ON. : official journal of the Balkan Union of Oncology, 18(1), 131-137 (2013-04-25)
As novel therapeutic agents relevant to colon cancer therapy are explored continuously, we tested 4 R2edda-type ligand precursors O,O'-dialkyl esters of (S,S)-ethylenediamine-N,N'-di-2-(4-methyl)pentanoic acid (L1.2HCl-L4.2HCl) and corresponding palladium(II) and platinum(II) complexes against the human colon cancer cell lines CaCo-2, SW480 and
Ling Li et al.
Nature chemistry, 5(7), 607-612 (2013-06-22)
The development of transition metal-catalysed cross-coupling reactions has greatly influenced the manner in which the synthesis of complex organic molecules is approached. A wide variety of methods are now available for the formation of C(sp(2))-C(sp(2)) bonds, and more recent work

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service