Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

639680

Sigma-Aldrich

trans-2-Hydroxy-β-nitrostyrene

97%

Synonym(s):

1-Hydroxy-2-(2-nitrovinyl)benzene, 2-(2-Hydroxyphenyl)nitroethene, 2-(2-Nitroethenyl)phenol, 2-Hydroxy-β-nitrostyrene, NSC 170698, trans-2-(2-Nitrovinyl)phenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH=CHNO2
CAS Number:
Molecular Weight:
165.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

functional group

amine
nitro

SMILES string

[H]\C(=C(\[H])[N+]([O-])=O)c1ccccc1O

InChI

1S/C8H7NO3/c10-8-4-2-1-3-7(8)5-6-9(11)12/h1-6,10H/b6-5+

InChI key

PMDYAIGGZBRBFX-AATRIKPKSA-N

Application

Reactant for:
  • Michael-acetalization reactions
  • Organocatalytic stereoselective Baylis-Hillman-type reactions with Hagemann′s esters
  • Catalyst-free aqueous-mediated conjugate addition reactions

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service