Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

632252

Sigma-Aldrich

Poly[4-(2,2-dicyanovinyl)-N-bis(hydroxyethyl)aniline-alt-(isophoronediisocyanate)]urethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((C14H13N3O2)(C12H19N2O2))n
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

mp

185 °C (lit.)

Application

NLO polymer

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service