Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

61800

Sigma-Aldrich

Lemongrass oil

natural, from Cymbopogon citratus D.C.

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103

biological source

Cymbopogon citratus D.C.

quality

natural

impurities

~75% citral

refractive index

n20/D 1.49

bp

224 °C (lit.)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

194.0 °F - closed cup

flash_point_c

90 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mansuang Wuthi-Udomlert et al.
The Southeast Asian journal of tropical medicine and public health, 42(2), 363-369 (2011-06-30)
Lemongrass (Cymbopogon citratus Stapf) has been used in cooking and in many traditional medicines; the essential oil contains citral as a major constituent. This study evaluated the antifungal activity of lemongrass oil against Malassezia furfur, an opportunistic yeast associated with
Shigeru Abe et al.
Mediators of inflammation, 13(1), 21-24 (2004-06-19)
In aromatherapy, essential oils are used as anti-inflammatory remedies, but experimental studies on their action mechanisms are very limited. To assess their anti-inflammatory activities, the effects of essential oils on neutrophil recruitment in mice were examined in vivo. The effect
V R Santhosh Kumar et al.
Asian Pacific journal of tropical medicine, 4(8), 658-660 (2011-09-15)
To evaluate the anti-hyperlipidemic activity of lemongrass oil against in dexamethasone induced hyperlipidemia in rats. Administration of dexamethasone was given at 10 mg/kg, sc. to the adult rats for 8 d induces hyperlipidemia characterized by marked increase in serum cholesterol
M Maizura et al.
Journal of food science, 72(6), C324-C330 (2007-11-13)
Edible films were prepared from a mixture of partially hydrolyzed sago starch and alginate (SA). Lemongrass oil (0.1% to 0.4%, v/w) and glycerol (0% and 20%, w/w) were incorporated in the films to act as natural antimicrobial agent and plasticizer
Jareerat Aiemsaard et al.
Research in veterinary science, 91(3), e31-e37 (2011-02-15)
The aims of this study were to investigate the antibacterial activity of lemongrass oil (LG) and its major components which were citral, geraniol and myrcene, against four strains of clinically isolated bovine mastitis pathogens, including Staphylococcus aureus, Streptococcus agalactiae, Bacillus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service