MilliporeSigma
  • The effect of lemongrass oil and its major components on clinical isolate mastitis pathogens and their mechanisms of action on Staphylococcus aureus DMST 4745.
The effect of lemongrass oil and its major components on clinical isolate mastitis pathogens and their mechanisms of action on Staphylococcus aureus DMST 4745.
Research in veterinary science (2011-02-15)
Jareerat Aiemsaard, Suneerat Aiumlamai, Chantana Aromdee, Suwimol Taweechaisupapong, Watcharee Khunkitti
ABSTRACT
The aims of this study were to investigate the antibacterial activity of lemongrass oil (LG) and its major components which were citral, geraniol and myrcene, against four strains of clinically isolated bovine mastitis pathogens, including Staphylococcus aureus, Streptococcus agalactiae, Bacillus cereus and Escherichia coli by the broth microdilution method, as well as their activity on S. aureus biofilm formation. Attempts to clarify their mechanisms of action by investigation of the effects on intracellular material leakage and morphological changes of S. aureus DMST 4745 were also made. The results demonstrate that S. agalactiae and B. cereus are more susceptible to LG, citral and geraniol than S. aureus and E. coli. Moreover, they also inhibit S. aureus biofilm formation and exhibit effective killing activities on preformed biofilms. The LG appears to have multiple targets in the bacterial cell, depending on concentration used as well as the amount of its components.