Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

609587

Sigma-Aldrich

Acrylonitrile-15N

≥98 atom % 15N, ≥99% (CP), contains hydroquinone as stabilizer

Synonym(s):

2-Propenenitrile-15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHC15N
CAS Number:
Molecular Weight:
54.06
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

≥98 atom % 15N

assay

≥99% (CP)

contains

hydroquinone as stabilizer

bp

77 °C (lit.)

mp

-83 °C (lit.)

density

0.830 g/mL at 25 °C

mass shift

M+1

storage temp.

2-8°C

SMILES string

C=CC#[15N]

InChI

1S/C3H3N/c1-2-3-4/h2H,1H2/i4+1

InChI key

NLHHRLWOUZZQLW-AZXPZELESA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

23.0 °F

flash_point_c

-4.99 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service