Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

591610

Sigma-Aldrich

Poly(pentabromophenyl methacrylate-co-glycidyl methacrylate)

glycidyl methacrylate 10 mol %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H5Br5O2)m(C7H10O3)n
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

composition

glycidyl methacrylate, 10 mol %

transition temp

Tg (DSC) 79 °C (onset)

solubility

chloroform: soluble

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

High refractive index, photosensitive polymer for optical waveguides.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service