Skip to Content
MilliporeSigma

574562

Vanadyl acetylacetonate

99.98% trace metals basis

Synonym(s):

VO(acac)2, Vanadium(IV)-oxy acetylacetonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
OV(C5H7O2)2
CAS Number:
Molecular Weight:
265.16
EC Number:
221-590-8
UNSPSC Code:
12352300
PubChem Substance ID:
Beilstein/REAXYS Number:
4138236
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99.98% trace metals basis

form

solid

mp

235 °C (dec.) (lit.)

SMILES string

CC(=O)\C=C(\C)O[V](=O)O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.O.V/c2*1-4(6)3-5(2)7;;/h2*3,6H,1-2H3;;/q;;;+2/p-2/b2*4-3-;;

InChI key

JFHJZWAQYMGNBE-SUKNRPLKSA-L

General description

Atomic number of base material: 23 Vanadium


Still not finding the right product?

Explore all of our products under Vanadyl acetylacetonate


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library