548510
Mercury(II) iodate
99%
Synonym(s):
Mercuric iodate, Mercury diiodate
About This Item
Recommended Products
assay
99%
form
powder
reaction suitability
reagent type: catalyst
core: mercury
SMILES string
I[Hg]I
InChI
1S/Hg.2HI/h;2*1H/q+2;;/p-2
InChI key
YFDLHELOZYVNJE-UHFFFAOYSA-L
Disclaimer

signalword
Danger
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2
Storage Class
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service