Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

546585

Sigma-Aldrich

Poly(9,9-dioctylfluorenyl-2,7-yleneethynylene)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

mp

>330 °C (lit.)

solubility

chloroform: soluble

fluorescence

λex 387 nm; λem 420 nm in chloroform

SMILES string

CCCCCCCCC1(CCCCCCCC)c2cc(ccc2-c3ccc(cc13)C#C)C#C.CCCCCCCCC4(CCCCCCCC)c5cc(ccc5-c6ccc(cc46)C#CC)C#C

General description

Poly(9,9-dioctylfluorenyl-2,7-yleneethynylene) (PFE) is a conjugating polymer that is a derivative of poly(fluoreneethynylene). It has high chemical stability with blue photo and electroluminescent properties which can be used in organic electronics.[1][2]

Application

Conducting polymer.
Exhibits blue photo- and electroluminescence.[3]
PFE is a conducting polymer which can be used in the fabrication of non-linear devices such as logic devices, modulators, and optical switches.[4] It may also be used in the formation of polymeric hybrids for ultraviolet photo-detector applications.[5][6]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Macromolecules, 33, 3961-3961 (2000)
Resonant third-order optical nonlinearities of chloroform solution and neat film of poly (9, 9-dioctylfluorenyl-2, 7-yleneethynylene)
Jin Z, et al.
Japanese Journal of Applied Physics, 45(8R), 6263-6263 (2006)
Poly (fluorenyleneethynylene) s by alkyne metathesis: Optical properties and aggregation behavior
Pschirer NG and Bunz UHF
Macromolecules, 33(11), 3961-3963 (2000)
Macromolecules, 33, 3961-3961 (2000)
Solution-processed polyfluorene: naphthalenediimide-N-doped TiO 2 hybrids for ultraviolet photodetector applications
Memisoglu G, et al.
Journal of Electronic Materials, 42(12), 3502-3511 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service