Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

544078

Sigma-Aldrich

Trifluoroacetonitrile

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CN
CAS Number:
Molecular Weight:
95.02
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Product 544078 is not currently sold in your country. Contact Technical Service

Quality Level

assay

99%

SMILES string

FC(F)(F)C#N

InChI

1S/C2F3N/c3-2(4,5)1-6

InChI key

SFFUEHODRAXXIA-UHFFFAOYSA-N

pictograms

Gas cylinderSkull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Inhalation - Press. Gas Compr. Gas

Storage Class

2A - Gases

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service