Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

542717

Sigma-Aldrich

2-Fluorobenzenesulfonamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4SO2NH2
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

162-166 °C (lit.)

SMILES string

NS(=O)(=O)c1ccccc1F

InChI

1S/C6H6FNO2S/c7-5-3-1-2-4-6(5)11(8,9)10/h1-4H,(H2,8,9,10)

InChI key

WFLBWYLZCQOPCA-UHFFFAOYSA-N

General description

2-Fluorobenzenesulfonamide can be synthesized by reacting 2-fluorobenzenesulfonyl chloride (in tetrahydrofuran) with 50% NH4OH.[1]

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L B Dugad et al.
Biochemistry, 28(9), 3955-3960 (1989-05-02)
Fluorine NMR has been used to examine complexes formed by 2-fluoro-, 3-fluoro-, and 2,5-difluorobenzenesulfonamide and human carbonic anhydrases I and II. The results show that all three inhibitors form complexes with both isozymes that have 2:1 inhibitor/enzyme stoichiometry. The fluorine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service