534528
Rink acid resin
50-100 mesh, extent of labeling: 0.6-0.8 mmol/g loading, 1 % cross-linked with divinylbenzene
Synonym(s):
(4-[(2,4-Dimethoxyphenyl)hydroxymethyl]phenol, polymer-bound, Rink acid-labile (hydroxymethyl)polystyrene
About This Item
Recommended Products
crosslinking
1 % cross-linked with divinylbenzene
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
extent of labeling
0.6-0.8 mmol/g loading
particle size
50-100 mesh
application(s)
peptide synthesis
functional group
alcohol
SMILES string
C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.COc3ccc(C(O)c4ccc(OCc5ccc(C=C)cc5)cc4)c(OC)c3
Looking for similar products? Visit Product Comparison Guide
Application
- Acid labile resin used in Fmoc-based solid phase peptide synthesis.
- Mild acidic cleavage conditions lead to the release of the peptide acid where fully protected peptides can be released if desired.
- Cleavage of the final protected peptide can be achieved in as little as 10% AcOH in DCM or 1% TFA in DCM.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service