Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

532819

Sigma-Aldrich

4-Bromo-3-methylbenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(CH3)CO2H
CAS Number:
Molecular Weight:
215.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

mp

212-216 °C (lit.)

SMILES string

Cc1cc(ccc1Br)C(O)=O

InChI

1S/C8H7BrO2/c1-5-4-6(8(10)11)2-3-7(5)9/h2-4H,1H3,(H,10,11)

InChI key

KWVXDZLVCISXIB-UHFFFAOYSA-N

General description

4-Bromo-3-methylbenzoic acid can be prepared by the hydrolysis of 4-bromo-3-methylbenzonitrile.[1]

Application

4-Bromo-3-methylbenzoic acid may be used in the preparation of following organic compounds:
  • 4-bromo-3-methylbenzoyl chloride[2]
  • 4-bromo-3-methylbenzoic acid tert-butyl ester[3]
  • 4-bromo-N-methoxy-3,N-dimethylbenzamide[4]
  • 2-(4-bromo-3-methylphenyl)-4,5-dihydro-4,4-dimethyloxazole[4]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

End group-functionalization and synthesis of block-copolythiophenes by modified nickel initiators.
Smeets A, et al.
Macromolecules, 44(15), 6017-6025 (2011)
Benzimidazole derivatives bearing substituted biphenyls as hepatitis C virus NS5B RNA-dependent RNA polymerase inhibitors: structure-activity relationship studies and identification of a potent and highly selective inhibitor JTK-109.
Hirashima S, et al.
Journal of Medicinal Chemistry, 49(15), 4721-4736 (2006)
Polycyclic systems. Part 19. Synthesis of 8-isobutyl-10-methyl-11 H-indeno [2, 1-a] phenanthrene (second diels hydrocarbon), a minor dehydrogenation product of cholesterol.
Nasipuri D, et al.
Journal of the Chemical Society. Perkin Transactions 1, 3034-3036 (1979)
Synthesis of polysaccharide derivatives bearing bromobenzoate pendants for use as chiral auxiliaries.
Ikai T, et al.
Reactive functional Polymers, 82, 52-57 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service