Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

53265

Sigma-Aldrich

Hexyltributylphosphonium bromide on polymer support

extent of labeling: ~0.5 mmol/g P loading

Synonym(s):

Polymer supported hexyltributylphosphonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

extent of labeling

~0.5 mmol/g P loading

matrix

polystyrene, crosslinked with 2% DVB

Other Notes

Very efficient phase-transfer catalyst. The catalyst is recovered by simple filtration. For a similar catalyst see[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Tundo et al.
Synthesis, 315-315 (1978)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service