Select a Size
Select a Size
About This Item
Recommended Products
assay
96%
optical activity
[α]20/D +189°, c = 1 in methanol
refractive index
n20/D 1.515 (lit.)
bp
267 °C (lit.)
density
1.073 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
OC[C@H]1CC2CCN1C[C@@H]2C=C
InChI
1S/C10H17NO/c1-2-8-6-11-4-3-9(8)5-10(11)7-12/h2,8-10,12H,1,3-7H2/t8-,9+,10+/m0/s1
InChI key
GAFZBOMPQVRGKU-IVZWLZJFSA-N
signalword
Warning
hcodes
pcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
235.0 °F - closed cup
flash_point_c
112.8 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service