Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

469076

Sigma-Aldrich

DAB-Am-16, Polypropylenimine hexadecaamine Dendrimer, Generation 3.0

volume 2,492 Å3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CH2N[(CH2)3N[(CH2)3N[(CH2)3NH2]2]2]2]2
CAS Number:
Molecular Weight:
1686.79
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

description

aminopropyl surface groups

mol wt

generation 3

feature

core type 1,4-diaminobutane core (4-carbon core)

no. Surface Groups

16

volume

2,492 Å3

refractive index

n20/D 1.515 (lit.)

viscosity

1.1 poise(50 °C)(lit.)

transition temp

Tg −90 °C

solubility

methanol and water: soluble

density

1.01 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

NCCCN(CCCN)CCCN(CCCN(CCCN)CCCN)CCCN(CCCCN(CCCN(CCCN(CCCN)CCCN)CCCN(CCCN)CCCN)CCCN(CCCN(CCCN)CCCN)CCCN(CCCN)CCCN)CCCN(CCCN(CCCN)CCCN)CCCN(CCCN)CCCN

InChI

1S/C88H208N30/c89-31-3-49-107(50-4-32-90)69-23-81-115(82-24-70-108(51-5-33-91)52-6-34-92)77-19-65-105(66-20-78-116(83-25-71-109(53-7-35-93)54-8-36-94)84-26-72-110(55-9-37-95)56-10-38-96)47-1-2-48-106(67-21-79-117(85-27-73-111(57-11-39-97)58-12-40-98)86-28-74-112(59-13-41-99)60-14-42-100)68-22-80-118(87-29-75-113(61-15-43-101)62-16-44-102)88-30-76-114(63-17-45-103)64-18-46-104/h1-104H2

InChI key

MVMZFIQOIDKSFP-UHFFFAOYSA-N

Physical form

Contains 16 primary amino groups.

Legal Information

DSM product

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service