Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

463140

Sigma-Aldrich

Diborane-d6

10% in helium, electronic grade

Synonym(s):

Diborane- d6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
B2D6
CAS Number:
Molecular Weight:
33.71
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

grade

electronic grade

form

gas (compressed)

concentration

10% in helium
9-11% (balance is helium)

bp

−92.5 °C (lit.)

mass shift

M+6

SMILES string

[2H]B([2H])[2H].[2H]B([2H])[2H]

InChI

1S/2BH3/h2*1H3/i2*1D3

InChI key

NTJHUEGHVSASEP-RVWRRERJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diborane gas is used primarily as a p-type dopant in the deposition of epitaxial and amorphous silicon thin films. The unique properties of diborane-d6 can result in increased device performance and reliability.

Recommended products

Stainless steel regulators Z527416 or Z527424 are recommended.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Gas 1A - Press. Gas Compr. Gas - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

2A - Gases

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service