Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

452939

Sigma-Aldrich

Trimethyl borate, azeotrope

70%, in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H9BO3
Molecular Weight:
103.91
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

70%

concentration

in methanol

refractive index

n20/D 1.3460

bp

53-58 °C

density

0.883 g/mL at 25 °C

SMILES string

COB(OC)OC

InChI

1S/C3H9BO3/c1-5-4(6-2)7-3/h1-3H3

InChI key

WRECIMRULFAWHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Rohm & Haas product

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

17.6 °F

flash_point_c

-8 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L V Angelova et al.
Soft matter, 11(25), 5060-5066 (2015-06-02)
Viscoelastic, gel-like, polymeric dispersions (HVPDs) can be prepared by crosslinking polyols with borax or boric acid in water under alkaline conditions. Rheologically similar HVPDs have been prepared in organic liquids containing no water or hydroxylic groups through crosslinking partially or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service