Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

439045

Supelco

Chromist aerosol propellant

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41115711
PubChem Substance ID:

vapor density

2.28 (vs air)

General description

CFC-free aerosol propellant. The Chromist attaches to the top of a standard 38-400 bottle filled with your reagent to deliver up to 16 oz of reagent in a fine mist spray for even coating. The unit minimizes waste and provides more consistent results for the chromatographer than in-house air.

Legal Information

Chromist is a trademark of Pall Gelman Sciences, Inc.

related product

Product No.
Description
Pricing

replaced by

Product No.
Description
Pricing

pictograms

FlameGas cylinder

signalword

Danger

hcodes

Hazard Classifications

Flam. Gas 1 - Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class

2A - Gases

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service