Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

432075

Sigma-Aldrich

Holmium(III) fluoride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Holmium trifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HoF3
CAS Number:
Molecular Weight:
221.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: holmium

SMILES string

F[Ho](F)F

InChI

1S/3FH.Ho/h3*1H;/q;;;+3/p-3

InChI key

FDIFPFNHNADKFC-UHFFFAOYSA-K

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral

supp_hazards

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service