Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

426776

Sigma-Aldrich

Poly(ethylene-co-acrylic acid) zinc salt

Salt of 426725

Synonym(s):

Acrylic acid-ethylene copolymer zinc salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

acrylic acid, 10 wt. %
acrylic acid, in zinc salt form, ~50%

hardness

1.1 dmm (ASTM D 5)

viscosity

5.5 poise(190 °C, Brookfield)(lit.)

acid number

40 mg KOH/g

transition temp

Tm 102 °C (DSC)

density

0.95 g/mL at 25 °C (lit.)

SMILES string

[Zn++].C=C.[O-]C(=O)C=C.[O-]C(=O)C=C

InChI

1S/C3H4O2.C2H4.Zn/c1-2-3(4)5;1-2;/h2H,1H2,(H,4,5);1-2H2;/p-1

Inchi Key

JJPWLZKJCBVHRG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Processing and performance additive. Promotes crystallization of PET. Assists dispersion of additives in plastics.

Features and Benefits

Forms reversible ionic clusters (crosslinks). Promotes adhesion to various substrates, tougher, more chemically resistant and more transparent than parent acid copolymer.

hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service