About This Item
Recommended Products
assay
98%
form
liquid
refractive index
n20/D 1.489 (lit.)
bp
180 °C/770 mmHg (lit.)
density
1.113 g/mL at 25 °C (lit.)
SMILES string
COC(=O)n1cccc1
InChI
1S/C6H7NO2/c1-9-6(8)7-4-2-3-5-7/h2-5H,1H3
InChI key
MORALDOSFHZOQS-UHFFFAOYSA-N
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Flam. Liq. 3
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
140.0 °F - closed cup
flash_point_c
60 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service