Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

420395

Sigma-Aldrich

(−)-Diisopropyl-O,O′-bis(trimethylsilyl)-D-tartrate

99%, ChiraSelect

Synonym(s):

(−)-O,O′-Bis(trimethylsilyl)-D-tartaric acid diisopropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH[OSi(CH3)3]CO2CH(CH3)2]2
CAS Number:
Molecular Weight:
378.61
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

product line

ChiraSelect

Quality Level

assay

99%

form

solid

optical activity

[α]20/D −57°, c = 1 in chloroform

refractive index

n20/D 1.428 (lit.)

bp

105-107 °C/1 mmHg (lit.)

density

0.972 g/mL at 25 °C (lit.)

SMILES string

CC(C)OC(=O)[C@@H](O[Si](C)(C)C)[C@H](O[Si](C)(C)C)C(=O)OC(C)C

InChI

1S/C16H34O6Si2/c1-11(2)19-15(17)13(21-23(5,6)7)14(22-24(8,9)10)16(18)20-12(3)4/h11-14H,1-10H3/t13-,14-/m0/s1

InChI key

WTAZEIUCUCCYIA-KBPBESRZSA-N

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service