Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

409731

Sigma-Aldrich

Methyl (S)-(−)-2-isocyanato-3-phenylpropionate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NCO)CO2CH3
CAS Number:
Molecular Weight:
205.21
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

assay

99%

optical activity

[α]20/D −80.0°, neat

refractive index

n20/D 1.513 (lit.)

bp

244 °C (lit.)

density

1.13 g/mL at 25 °C (lit.)

SMILES string

COC(=O)[C@H](Cc1ccccc1)N=C=O

InChI

1S/C11H11NO3/c1-15-11(14)10(12-8-13)7-9-5-3-2-4-6-9/h2-6,10H,7H2,1H3/t10-/m0/s1

InChI key

JOTMMIYKEOOTNZ-JTQLQIEISA-N

Application

This compound has been employed in the preparation of dipeptide analogs of angiotensinogen[1] and Schiff base derivatives for the inhibition of thermolysin.[2]

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

116.1 °F - closed cup

flash_point_c

46.7 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bates, S.R.E. et al.
J. Chem. Res. Synop., 48-48 (1993)
J J Plattner et al.
Journal of medicinal chemistry, 31(12), 2277-2288 (1988-12-01)
A series of renin inhibitors have been prepared and evaluated for their susceptibility to cleavage by the serine protease chymotrypsin. The compounds were designed by consideration of the structural requirements in the active-site region of renin and chymotrypsin. By systematic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service