Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40301

Sigma-Aldrich

2-Propanol

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein/REAXYS Number:
635639
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
bp:
82 °C (lit.)
vapor pressure:
33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

form

liquid

CofA

specification on request

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

color

colorless

refractive index

n20/D 1.377 (lit.)
n/D

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

solubility

water: soluble (completely)

density

0.785 g/mL at 25 °C (lit.)

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

53.6 °F - closed cup

flash_point_c

12.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Peter C Grayson et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(7), 1922-1932 (2015-04-22)
To discover biomarkers involved in the pathophysiology of antineutrophil cytoplasmic antibody-associated vasculitis (AAV) and to determine whether low-density granulocytes (LDGs) contribute to gene expression signatures in AAV. The source of clinical data and linked biologic specimens was a randomized controlled
Jan Rybniker et al.
Nature communications, 6, 7659-7659 (2015-07-15)
Better antibiotics capable of killing multi-drug-resistant Mycobacterium tuberculosis are urgently needed. Despite extensive drug discovery efforts, only a few promising candidates are on the horizon and alternative screening protocols are required. Here, by testing a panel of FDA-approved drugs in
H Dolatshad et al.
Leukemia, 29(5), 1092-1103 (2014-11-28)
The splicing factor SF3B1 is the most commonly mutated gene in the myelodysplastic syndrome (MDS), particularly in patients with refractory anemia with ring sideroblasts (RARS). We investigated the functional effects of SF3B1 disruption in myeloid cell lines: SF3B1 knockdown resulted
Thomas W Winkler et al.
PLoS genetics, 11(10), e1005378-e1005378 (2015-10-02)
Genome-wide association studies (GWAS) have identified more than 100 genetic variants contributing to BMI, a measure of body size, or waist-to-hip ratio (adjusted for BMI, WHRadjBMI), a measure of body shape. Body size and shape change as people grow older
Emilie Montastier et al.
PLoS computational biology, 11(1), e1004047-e1004047 (2015-01-16)
Nutrigenomics investigates relationships between nutrients and all genome-encoded molecular entities. This holistic approach requires systems biology to scrutinize the effects of diet on tissue biology. To decipher the adipose tissue (AT) response to diet induced weight changes we focused on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service