Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

392243

Sigma-Aldrich

1-Formylpiperazine

technical grade, 85%

Synonym(s):

Piperazine-1-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10N2O
CAS Number:
Molecular Weight:
114.15
Beilstein/REAXYS Number:
110295
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

assay

85%

form

liquid

density

1.107 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)N1CCNCC1

InChI

1S/C5H10N2O/c8-5-7-3-1-6-2-4-7/h5-6H,1-4H2

Inchi Key

MSSDTZLYNMFTKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Formylpiperazine, a secondary alicyclic amine[1] is a piperazine derivative. Its synthesis has been reported.[2] The kinetics of its reaction with O-aryl S-aryl dithiocarbonates has been studied.[3] The IR and Raman spectra of 1-formylpiperazine have been investigated.[4]

Application

1-Formylpiperazine is sutable for use as a test compound in the kinetic study of the reaction of 4-nitrophenyl 2-thiophenecarboxylate with secondary alicyclic amines.[1]

Pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

215.6 °F - closed cup

flash_point_c

102 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactions of O-aryl S-aryl dithiocarbonates with secondary alicyclic amines in aqueous ethanol. Kinetics and mechanism.
Castro EA, et al.
Journal of Physical Organic Chemistry, 24(6), 466-473 (2011)
Kinetics and mechanism for the reaction of 4-nitrophenyl 2-thiophenecarboxylate with secondary alicyclic amines.
Um IH, et al.
Bull. Korean Chem. Soc., 23(3), 381-384 (2002)
Gürkan Keşan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 118, 1113-1120 (2013-10-29)
Infrared and Raman spectra of 1-formylpiperazine (1-fp) have been recorded in the region of 4000-50 cm(-1). The conformational analysis, optimized geometric parameters, normal mode frequencies and corresponding vibrational assignments of 1-fp (C5H10N2O) have been examined by means of Becke-3-Lee-Yang-Parr (B3LYP)
1-Formylpiperazine and related compounds.
Horrom BW, et al.
Journal of the American Chemical Society, 77(3), 753-754 (1955)
Gu-cai Li et al.
Die Pharmazie, 70(8), 511-514 (2015-09-19)
Five indolin-2-one derivatives bearing piperazinylbutyl side chains attached to the amide nitrogen were synthesized from 2-indolinone. 1-(4-Bromobutyl)-indolin-2-one was reacted with 1-piperazinecarboxaldehyde to form 1-(4-(4-formyl-1-piperazinyl)butyl)indolin-2-one (2). In the presence of H2SO4, the aldehyde moiety was removed from 1-(4-(4-formyl-1-piperazinyl)butyl)indolin-2-one and then 1-(4-(1-piperazinyl)butyl)indolin-2-one

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service