391344
(S)-(+)-2-Oxo-4-phenyl-3-oxazolidineacetic acid
98%
About This Item
Recommended Products
assay
98%
optical activity
[α]28/D +165°, c = 2 in chloroform
mp
103-105 °C (lit.)
SMILES string
OC(=O)CN1[C@H](COC1=O)c2ccccc2
InChI
1S/C11H11NO4/c13-10(14)6-12-9(7-16-11(12)15)8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,13,14)/t9-/m1/s1
Inchi Key
PUKMRNJLFMISMT-SECBINFHSA-N
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service