Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

380253

Sigma-Aldrich

Ethyl pipecolinate hydrochloride

97%

Synonym(s):

Ethyl 2-piperidinecarboxylate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO2 · HCl
CAS Number:
Molecular Weight:
193.67
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

211-213 °C (lit.)

SMILES string

Cl[H].CCOC(=O)C1CCCCN1

InChI

1S/C8H15NO2.ClH/c1-2-11-8(10)7-5-3-4-6-9-7;/h7,9H,2-6H2,1H3;1H

InChI key

KGAWPIXNSIYQPC-UHFFFAOYSA-N

General description

Ethyl pipecolinate hydrochloride is a heterocyclic building block. It is reported to participate in the total synthesis of the (+)-epiquinamide, a quinolizidine alkaloid.[1]

Application

Ethyl pipecolinate hydrochloride (Ethyl 2-piperidinecarboxylate hydrochloride) may be used in the preparation of:
  • 1,4-diazabicyclo[4.4.0]decane[2]
  • small molecule inhibitors of Legionella MIP protein[3]
  • antiinflammatory and analgesic molecules[4]
  • anti-amnesic agents[5]

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christina Juli et al.
Journal of medicinal chemistry, 54(1), 277-283 (2010-12-15)
The macrophage infectivity potentiator (MIP) protein is a major virulence factor of Legionella pneumophila, the causative agent of Legionnaires' disease. MIP belongs to the FK506-binding proteins (FKBP) and is necessary for optimal intracellular survival and lung tissue dissemination of L.
A concise synthesis of (?) and a total synthesis of (+)-epiquinamide.
Tong STA and Barker D.
Tetrahedron Letters, 47(29), 5017-5020 (2006)
Syntheses of 1, 4-Diazabicyclo [4.4. 0] decanes, 1, 4-Diazabicyclo [4.3. 0] nonanes and 1, 8-Diazabicyclo [4.3. 0] nonanes.
Freed ME and Day AR.
J. Org. React., 25(12), 2108-2113 (1960)
G. A. Gevorgyan, et al.,
Khim.-Farm. Zh., 37, 152-154 (2003)
S Zhao et al.
Bioorganic & medicinal chemistry, 7(8), 1637-1646 (1999-09-11)
Tacrine, one of the drugs available for Alzheimer's disease based on the cholinergic approach, suffers from considerable toxicity. Many analogues of tacrine have been prepared which retain the pharmacologically rich aminopyridine or aminoquinoline motifs. The current research was undertaken to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service