Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

364665

Sigma-Aldrich

Ethylmagnesium bromide solution

1.0 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2MgBr
CAS Number:
Molecular Weight:
133.27
Beilstein/REAXYS Number:
3587203
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in THF

density

1.01 g/mL at 25 °C

SMILES string

CC[Mg]Br

InChI

1S/C2H5.BrH.Mg/c1-2;;/h1H2,2H3;1H;/q;;+1/p-1

InChI key

TWTWFMUQSOFTRN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Storage below 25°C may cause formation of crystalline magnesium salts. Moving container to a warm location and occasional gentle swirling should redissolve the solid.

Recommended products

Angled septum-inlet adapter Z118206 or straight septum-inlet adapter, 6mm I.D. inlet, Z118141 or straight septum-inlet adapter, 13mm I.D. inlet, Z118192 is recommended.

septum inlet adapter

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

target_organs

Respiratory system

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 1

flash_point_f

23.0 °F - closed cup

flash_point_c

-5 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Nishiyama et al.
Chemical & pharmaceutical bulletin, 48(12), 1999-2002 (2001-01-06)
The direct alpha-bromination of various ketones using trifluoromethanesulfonic anhydride and Grignard reagent or magnesium bromide in ether gave the corresponding alpha-bromo ketones in moderate to good yields under mild reaction conditions.
F Turon et al.
Lipids, 37(8), 817-821 (2002-10-10)
An analytical procedure was developed for regiodistribution analysis of TAG using alpha-MAG prepared by an ethyl magnesium bromide deacylation. In the present communication, the deacylation procedure is shown to lead to representative alpha-MAG, allowing the composition of the native TAG
R G Xie et al.
Steroids, 55(11), 488-490 (1990-11-01)
The reaction of ethyl magnesium bromide and 17 alpha-ethynylestradiol with formaldehyde in the presence of triethyl phosphate or hexamethylphosphoramide gave the 2- and 4-formyl-17 alpha-ethynylestradiol in high yield. Treatment of the formyl derivatives with an alkaline solution of hydrogen peroxide
Janne E Tønder et al.
Organic & biomolecular chemistry, 2(10), 1447-1455 (2004-05-12)
Copper mediated allylic substitutions and conjugate additions to geranyl, cinnamyl and allylic indole compounds have been investigated with the aim of finding a method for the creation of the all-carbon quaternary centre present in the natural product lyngbyatoxin A. Reaction
M Wältermann et al.
Microbiology (Reading, England), 146 ( Pt 5), 1143-1149 (2000-06-01)
The triacylglycerol (TAG)-accumulating, hydrocarbon-degrading bacterium Rhodococcus opacus strain PD630 and chemically induced storage-deficient mutants derived from this strain were investigated for their capability to accumulate storage lipids in the cytoplasm during cultivation under nitrogen-limiting conditions. Acylglycerols were analysed by matrix-associated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service