About This Item
Recommended Products
Quality Level
assay
97%
form
liquid
refractive index
n20/D 1.451 (lit.)
density
1.319 g/mL at 25 °C (lit.)
SMILES string
NCc1cc(F)cc(c1)C(F)(F)F
InChI
1S/C8H7F4N/c9-7-2-5(4-13)1-6(3-7)8(10,11)12/h1-3H,4,13H2
InChI key
ADHUUMWSWNPEMZ-UHFFFAOYSA-N
General description
Application
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
170.6 °F - closed cup
flash_point_c
77 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service