Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

333565

Sigma-Aldrich

Cyclobutyl chloride

97%

Synonym(s):

Chlorocyclobutane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7Cl
CAS Number:
Molecular Weight:
90.55
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

83 °C (lit.)

density

0.991 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

ClC1CCC1

InChI

1S/C4H7Cl/c5-4-2-1-3-4/h4H,1-3H2

InChI key

STJYMUBZVMSMBP-UHFFFAOYSA-N

General description

Cyclobutyl chloride is formed by rearrangements concerted with fragmentation of cyclopropylmethoxychlorocarbene and cyclobutoxychlorocarbene.[1] The thermal decomposition of cyclobutyl chloride was investigated over the temperature range of 892-1150K using very low-pressure pyrolysis technique.[2]

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

15.8 °F - closed cup

flash_point_c

-9 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Competitive unimolecular reactions at low pressures. The pyrolysis of cyclobutyl chloride.
King KD, et al.
International Journal of Chemical Kinetics, 11(1), 11-21 (1979)
Robert A Moss et al.
Journal of the American Chemical Society, 126(27), 8466-8476 (2004-07-09)
Fragmentations of cyclopropylmethoxychlorocarbene (6) and cyclobutoxychlorocarbene (10) lead to rearrangments that afford mixtures of cyclopropylmethyl chloride (7), cyclobutyl chloride (8), and 3-butenyl chloride (9). Isotopic substitution studies show that these rearrangments are accompanied by partial exchange of the methylene groups

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service