Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

302139

Sigma-Aldrich

Potassium triphenylborohydride solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KB(C6H5)3H
CAS Number:
Molecular Weight:
282.23
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

form

liquid

reaction suitability

reagent type: reductant

concentration

0.5 M in THF

density

0.94 g/mL at 25 °C

SMILES string

[K+].c1ccc(cc1)[BH-](c2ccccc2)c3ccccc3

InChI

1S/C18H16B.K/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;/h1-15,19H;/q-1;+1

InChI key

RHWPQWOBMSRXHC-UHFFFAOYSA-N

Application

  • Reducing agent
  • Reagent for the reduction of a,ß-unsaturated carboxylic acid derivatives

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3 - Water-react 1

target_organs

Respiratory system

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

-0.4 °F - closed cup

flash_point_c

-18 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service