Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

279277

Sigma-Aldrich

N-(Hydroxymethyl)nicotinamide

98%

Synonym(s):

3-Pyridinecarboxylic acid N-hydroxymethylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N2O2
CAS Number:
Molecular Weight:
152.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

mp

152-154 °C (lit.)

functional group

amide
hydroxyl

SMILES string

OCNC(=O)c1cccnc1

InChI

1S/C7H8N2O2/c10-5-9-7(11)6-2-1-3-8-4-6/h1-4,10H,5H2,(H,9,11)

Inchi Key

JRFKIOFLCXKVOT-UHFFFAOYSA-N

General description

A quantitative determination of N-(hydroxymethyl)nicotinamide in tablets and basic solutions along with nicotinic acid by thin-layer chromatography-densitometric method has been reported[1]. N-(Hydroxymethyl)nicotinamide is an antimicrobial agent[2].

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Urszula Hubicka et al.
Journal of AOAC International, 91(5), 1186-1190 (2008-11-05)
A thin-layer chromatography (TLC)-densitometric method was developed to determine N-(hydroxymethyl)nicotinamide in tablets and basic solutions along with nicotinic acid. Analysis was performed on silica gel F254 plates using chloroform-ethanol (2 + 3, v/v) mobile phase. The densitometric observations were made
H K Zhang et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 27(8), 629-631 (1992-01-01)
N-Hydroxymethyl-nicotinamide, an antimicrobic agent was prepared in good yield by treating nicotinamide with excess of formaldehyde in the presence of potassium carbonate. The condensation reaction was studied under different conditions, changing the amount of formaldehyde, with different kinds of bases

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service