Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

278750

Sigma-Aldrich

(1R,2S,5R)-(−)-Menthyl (S)-p-toluenesulfinate

98%

Synonym(s):

(−)-(1R)-Menthyl (S)-p-toluenesulfinate, (S)-(−)-Menthyl p-toluenesulfinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26O2S
CAS Number:
Molecular Weight:
294.45
Beilstein/REAXYS Number:
3207468
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

optical activity

[α]20/D −195°, c = 2 in acetone

mp

102-104 °C (lit.)

functional group

sulfinic acid

storage temp.

−20°C

SMILES string

C[C@@H]1CC[C@@H](C(C)C)[C@@H](C1)OS(=O)c2ccc(C)cc2

InChI

1S/C17H26O2S/c1-12(2)16-10-7-14(4)11-17(16)19-20(18)15-8-5-13(3)6-9-15/h5-6,8-9,12,14,16-17H,7,10-11H2,1-4H3/t14-,16+,17-,20+/m1/s1

InChI key

NQICGNSARVCSGJ-ASKNOMKYSA-N

General description

(1R,2S,5R)-(-)-Menthyl (S)-p-toluenesulfinate is mainly used for the introduction of p-toluenesulfinyl group in asymmetric synthesis.[1]

Application

(1R,2S,5R)-(-)-Menthyl (S)-p-toluenesulfinate can react with:
  • metal ketimines to form enantiopure sulfinimines[1]
  • p-(methylthio)benzaldehyde to form (S)-(-)-N-(4-methylthiobenzylidene)-p-toluenesulfinimine[2]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric synthesis and properties of sulfinimines (thiooxime S-oxides).
Davis FA, et al.
The Journal of Organic Chemistry, 62(8), 2555-2563 (1997)
Asymmetric synthesis of the antibiotic (+)-thiamphenicol using cis-N-(p-toluenesulfinyl) aziridine 2-carboxylic acids
Davis FA and Zhou P.
Tetrahedron Letters, 35(41), 7525-7528 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service