Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

277916

Sigma-Aldrich

Rhodium

wire, diam. 1.0 mm, 99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Rh
CAS Number:
Molecular Weight:
102.91
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

99.9% trace metals basis

form

wire

resistivity

4.33 μΩ-cm, 20°C

diam.

1.0 mm

bp

3727 °C (lit.)

mp

1966 °C (lit.)

density

12.41 g/cm3 (lit.)

SMILES string

[Rh]

InChI

1S/Rh

InChI key

MHOVAHRLVXNVSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

200 mg = 2 cm; 1 g = 10 cm

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon Duttwyler et al.
Science (New York, N.Y.), 339(6120), 678-682 (2013-02-09)
Piperidines are prevalent in natural products and pharmaceutical agents and are important synthetic targets for drug discovery and development. We report on a methodology that provides highly substituted piperidine derivatives with regiochemistry selectively tunable by varying the strength of acid
Stepan Chuprakov et al.
Journal of the American Chemical Society, 135(12), 4652-4655 (2013-03-13)
Readily available 1-mesyl-1,2,3-triazoles are efficiently converted into a variety of imidazolones and thiazoles by Rh(II)-catalyzed denitrogenative reactions with isocyanates and isothiocyanates, respectively. The proposed triazole-diazoimine equilibrium results in the formation of highly reactive azavinyl metal-carbenes, which react with heterocumulenes causing
Juliane Keilitz et al.
Organic letters, 15(5), 1148-1151 (2013-02-21)
A new enantioselective rhodium-catalyzed domino reaction is described that gives access to fused heterocycles by desymmetrization of alkyne-tethered cyclohexadienones. Two new C-C bonds and two stereocenters are formed in one step with good enantioselectivity. In contrast to prior reports, it
Xian-Ying Shi et al.
Organic letters, 15(7), 1476-1479 (2013-03-09)
A novel rhodium(III)-catalyzed direct functionalization of the ortho-C-H bond of aromatic ketone derivatives and an intramolecular cyclization sequence produced indene derivatives in moderate to good yields. This cascade cyclization involves a conjugate addition of α,β-unsaturated ketone and subsequent aldol condensation.
P Andrew Evans et al.
Organic letters, 15(8), 1798-1801 (2013-04-02)
A concise and highly convergent three-step total synthesis of the lactarane natural product, pyrovellerolactone, is described. The key step involves a regio- and diastereoselective rhodium-catalyzed [(3 + 2) + 2] carbocyclization of an alkenylidenecyclopropane with a 4-hydroxybut-2-ynoate followed by an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service