Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

257109

Sigma-Aldrich

Methylmagnesium iodide solution

3.0 M in diethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3MgI
CAS Number:
Molecular Weight:
166.24
Beilstein/REAXYS Number:
1209226
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

reaction suitability

reaction type: Grignard Reaction

concentration

3.0 M in diethyl ether

density

1.261 g/mL at 25 °C

SMILES string

C[Mg]I

InChI

1S/CH3.HI.Mg/h1H3;1H;/q;;+1/p-1

InChI key

AUPXBVDHVRZMIB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Recommended products

Angled septum-inlet adapter Z118206 or straight septum-inlet adapter, 6mm I.D. inlet, Z118141 or straight septum-inlet adapter, 13mm I.D. inlet, Z118192 is recommended.

septum inlet adapter

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

target_organs

Central nervous system

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

-40.0 °F - closed cup

flash_point_c

-40 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service