About This Item
Recommended Products
reaction suitability
reaction type: Grignard Reaction
concentration
2.0 M in THF
density
0.995 g/mL at 25 °C
SMILES string
Cl[Mg]CC=C
InChI
1S/C3H5.ClH.Mg/c1-3-2;;/h3H,1-2H2;1H;/q;;+1/p-1
InChI key
PLYLAFITHJUEGX-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Packaging
Compatible with the following:
- Aldrich® Sure/Pac™ station for liquefied gases Z566446
- PTFE Sealing tape Z104388 or Z221880
- Straight septum-inlet adapter Z118141 with septa Z565687 or Z565695
Legal Information
also commonly purchased with this product
recommended
septum inlet adapter
signalword
Danger
Hazard Classifications
Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1
target_organs
Central nervous system, Respiratory system
supp_hazards
Storage Class
4.3 - Hazardous materials which set free flammable gases upon contact with water
wgk_germany
WGK 1
flash_point_f
1.4 °F - closed cup
flash_point_c
-17 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service