Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

250732

Sigma-Aldrich

1,5-Dithiacyclooctan-3-ol

technical grade, 90%

Synonym(s):

1,5-Dithiocan-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12OS2
CAS Number:
Molecular Weight:
164.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

assay

90%

refractive index

n20/D 1.597 (lit.)

bp

147 °C/2 mmHg (lit.)

density

1.272 g/mL at 25 °C (lit.)

functional group

hydroxyl
thioether

SMILES string

OC1CSCCCSC1

InChI

1S/C6H12OS2/c7-6-4-8-2-1-3-9-5-6/h6-7H,1-5H2

InChI key

WUMSAPPXLGVSSD-UHFFFAOYSA-N

General description

1,5-Dithiacyclooctan-3-ol forms thioether complexes with lead (II)[1].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cadmium(II), bismuth(III), lead(II) and thallium(I) crown thioether chemistry: synthesis and crystal structures of [(CdI2) 2 ([24] aneS8)],[(BiCl3) 2 ([24] aneS8)],[Pb2 ([28] aneS8)][ClO4]4 and [Tl ([24] aneS8)] PF6
Blake AJ, et al.
J. Chem. Soc., Dalton Trans., 23, 3961-3968 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service