Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

225495

Sigma-Aldrich

Disodium tetracarbonylferrate dioxane complex

Synonym(s):

Collman’s Reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2Fe(CO)4 · 1.5C4H8O2
CAS Number:
Molecular Weight:
346.02
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: iron
reagent type: catalyst

SMILES string

[Fe--].[Fe--].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Na]C#[O+].[Na]C#[O+].[Na]C#[O+].[Na]C#[O+].C1COCCO1.C2COCCO2.C3COCCO3

InChI

1S/3C4H8O2.8CO.2Fe.4Na/c3*1-2-6-4-3-5-1;8*1-2;;;;;;/h3*1-4H2;;;;;;;;;;;;;;/q;;;;;;;4*+1;2*-2;;;;

InChI key

XEXTUUGKJXEJOB-UHFFFAOYSA-N

Application

Carbonylating agent for halides to form aldehydes andketones; hydroacylates Michael acceptors; also used as reducing agent; catalyst for dismutation of aromatic aldehydesto esters

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Carc. 2 - Pyr. Sol. 1 - Water-react 2

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Accounts of Chemical Research, 8, 342-342 (1975)
McMurray, J. E.; Andrus, A.
Tetrahedron Letters, 21, 4687-4687 (1980)
Aldrichimica Acta, 19, 69-69 (1986)
Accounts of Chemical Research, 1, 136-136 (1968)
Yamashita, M.; Tashika, H. et al.
Bulletin of the Chemical Society of Japan, 65, 1257-1257 (1992)

Questions

1–3 of 3 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is the physical form of Product 225495, Disodium tetracarbonylferrate dioxane complex?

    1 answer
    1. This material is reported to be a solid, powder and/or chunks.

      Helpful?

  3. Is Product 225495, Disodium tetracarbonylferrate dioxane complex, the same as Collman's Reagent?

    1 answer
    1. Yes, Collman's Reagent is a synonym for this product. The compound is described by James P. Collman in the following reference: Acc. Chem. Res, 8(10), 342-347(1975).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service