Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

217166

Sigma-Aldrich

1-Methoxy-1,3-cyclohexadiene

technical grade, 65%

Synonym(s):

2,3-Dihydroanisole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O
CAS Number:
Molecular Weight:
110.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

concentration

65%

refractive index

n20/D 1.488 (lit.)

bp

40 °C/15 mmHg (lit.)

density

0.929 g/mL at 25 °C (lit.)

SMILES string

COC1=CC=CCC1

InChI

1S/C7H10O/c1-8-7-5-3-2-4-6-7/h2-3,5H,4,6H2,1H3

InChI key

LNRUJSUKKBJFOM-UHFFFAOYSA-N

General description

1-Methoxy-1,3-cyclohexadiene is a useful Diels-Alder diene employed in Diels-Alder cycloadditions.[1] It undergoes Diels-Alder reaction with alkyne to yield anisole derivative.[2]

application

1-Methoxy-1,3-cyclohexadiene was used in the following studies:
  • Synthesis of tetracyclic polyhydroxyquinones via Diels-Alder reaction.[3]
  • Synthesis of tetra-ortho-substituted biaryls via Diels-Alder reaction.[1]
  • Synthesis of optically active form of dihydroisocoumarin fragment of the gastroprotective natural product AI-77-B, via regiospecific Diels-Alder reaction with an acetylenic ester derivative.[4]

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

80.6 °F - closed cup

flash_point_c

27 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bradley O Ashburn et al.
Journal of the American Chemical Society, 129(29), 9109-9116 (2007-07-03)
The application of the Diels-Alder approach to biaryls (DAB) is described for the synthesis of tetra-ortho-substituted biaryl compounds containing orthogonally functionalized substituents. The syntheses of phosphorus-containing, disubstituted alkynes and carbonyl-containing, disubstituted alkynes were accomplished in two to three steps from
Stereoselective synthesis of dihydroisocoumarin moiety of microbial agent AI-77-B: a Diels-Alder based strategy.
Ghosh AK and Cappiello J.
Tetrahedron Letters, 39(48), 8803-8806 (1998)
M. Carmen Carreño et al.
The Journal of organic chemistry, 61(18), 6136-6138 (1996-09-06)
The synthesis of tetracyclic polyhydroxyquinones 5a and 5b was achieved through a sequence involving two Diels-Alder reactions with 1-methoxy-1,3-cyclohexadiene: the first with 2-(p-tolylthio)naphthazarin and the second on the resulting tricyclic derivative previously transformed into a (p-tolylsulfinyl)naphthazarin. The success of this
Bradley O Ashburn et al.
Organic & biomolecular chemistry, 6(2), 255-257 (2008-01-05)
A series of nitrophenyl acetylenes were evaluated for their utility in a cycloaddition-cycloreversion approach to polysubstituted biaryls. ortho-Nitrophenyl acetylene consistently provided the target biaryls in superior yields as compared to the meta and para nitro substituted variants.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service