Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

197157

Sigma-Aldrich

4-Morpholinoaniline

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O
CAS Number:
Molecular Weight:
178.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥98%

form

solid

mp

132-135 °C (lit.)

functional group

ether

SMILES string

Nc1ccc(cc1)N2CCOCC2

InChI

1S/C10H14N2O/c11-9-1-3-10(4-2-9)12-5-7-13-8-6-12/h1-4H,5-8,11H2

InChI key

PHNDZBFLOPIMSM-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A green approach for the electrochemical synthesis of 4-morpholino-2-(arylsulfonyl) benzenamines.
Nematollahi D and Esmaili R.
Tetrahedron Letters, 51(37), 4862-4865 (2010)
Roya Esmaili et al.
The Journal of organic chemistry, 78(10), 5018-5021 (2013-05-01)
Electrochemical synthesis of two new disulfides of 2-(benzo[d]thiazol(or oxazol)-2-ylamino)-5-morpholinobenzene thiols was carried out via the electrooxidation of 4-morpholinoaniline in the presence of 2-mercaptobenzothiazole and 2-mercaptobenzoxazole. Our results indicate that electrogenerated p-quinonediimine participated in a Michael-type addition reaction with 2-SH-benzazoles and
K Dhahagani et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 117, 87-94 (2013-08-30)
Metal(II) chelates of Schiff bases derived from the condensation of 4-morpholinoaniline with substituted salicylaldehyde have been prepared and characterized by (1)H NMR, IR, electronic, EPR, and magnetic measurement studies. The complexes are of the type M(X-MPMP)2 [where M=Cu(II), Co(II)), Zn(II)
Carey L Horchler et al.
Bioorganic & medicinal chemistry, 15(2), 939-950 (2006-11-28)
Multiparallel amenable syntheses of 6-methoxy-8-amino-4-oxo-1,4-dihydroquinoline-2-carboxylic acid-(4-morpholin-4-yl-phenyl)amides (I) and 4-amino-6-methoxy-8-(4-methyl-piperazin-1-yl)-quinoline-2-carboxylic acid (4-morpholin-4-yl-phenyl)amides (II) which facilitate late-stage diversification at the 8-position of (I) and at the 4- and 8-positions of (II) are described. The resulting novel series were determined to contain potent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service