Skip to Content
Merck
All Photos(1)

Key Documents

138045

Sigma-Aldrich

L-Tyrosine hydrazide

98%, suitable for bioconjugation

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2CH(NH2)CONHNH2
CAS Number:
Molecular Weight:
195.22
Beilstein:
2808635
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Product Name

L-Tyrosine hydrazide, 98%

Assay

98%

form

powder

optical activity

[α]20/D +76°, c = 4.2 in 1 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

technique(s)

bioconjugation: suitable

color

white to faint tan

mp

196-198 °C (lit.)

application(s)

peptide synthesis

SMILES string

NNC(=O)[C@@H](N)Cc1ccc(O)cc1

InChI

1S/C9H13N3O2/c10-8(9(14)12-11)5-6-1-3-7(13)4-2-6/h1-4,8,13H,5,10-11H2,(H,12,14)/t8-/m0/s1

InChI key

MWIXENPCUPDSOS-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Excellent reagent for the resolution of amino acids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xianfeng Gu et al.
Journal of medicinal chemistry, 49(25), 7532-7539 (2006-12-08)
Protein tyrosine kinases use two Mg(2+) ions as cofactors in catalysis, one as the ATP-Mg complex (M1) and the other as an essential activator (M2). The M2-binding site has high affinity for transition metal cations such as cobalt and zinc.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service