Principaux documents
Z719471
Sorenson low binding aerosol barrier tips
MultiGuard, with graduation lines, maximum volume 200 μL
Synonyme(s) :
aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, low bind tip, pipet tip
About This Item
Code UNSPSC :
41121601
Nomenclature NACRES :
NB.01
Matériaux
colorless
natural tip (low binding)
polypropylene
Stérilité
sterile
sterile
Caractéristiques
DNase free
RNase free
barrier
Conditionnement
pack of 96 ea (Racked)
Fabricant/nom de marque
Sorenson 17370T
Volume maximum
200 μL
Type de liaison
low binding surface
Description générale
Tips feature a new surface technology designed for quantitative assays in which sample binding can significantly alter results or for critical small volume protocols that require a high degree of accuracy. Ideal for molecular biology applications, such as PCR. Inert hydrophobic barrier prevents pipettor contamination.
Caractéristiques et avantages
  • Suitable for DNA amplification and radioisotope handling
  • Over-pipetting does not contaminate valuable samples
  • Universal fit on research-grade pipettors
  • Filter devoid of leachable additives
  • Small micron pore sizes, <20 μ
  • Hydrophobic surface to lower surface tension
  • Minimal sample binding
  • Enhanced accuracy and precision
  • Additives-free such as silicone
  • Eliminates leaching and subsequent sample degradation
Informations légales
Sorenson is a trademark of Sorenson BioScience, Inc.