Principaux documents
MTCP0800TL
MTC Bio PCR Tube Strip Opener
Synonyme(s) :
PCR strip opener, cell sieve, flow cytometry tube, tube opender
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.15
Conditionnement
pack of 2 ea
Fabricant/nom de marque
MTC Bio P0800-TL
Description générale
Tool for opening/closing PCR strips, pk/2
Caractéristiques et avantages
  • RNase, DNase, DNA, and PCR inhibitor free
  • 0.2mL or Low Profile 0.1mL options
  • Ultra-thin, uniform wall thickness
  • White and frosted tubes for enhanced qPCR results
Informations légales
MTC is a trademark of MTC Bio