Principaux documents
Z247758
Coors high-alumina crucible cover
diam. 59 mm
About This Item
Code UNSPSC :
41122200
Nomenclature NACRES :
NB.43
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Coorstek 65613
Diamètre
59 mm
Description générale
Covers may not fit exactly on crucibles due to unavoidable wall thickness variations within the crucible.
For Z247642
Informations légales
Coors is a trademark of Coorstek, Inc.
Les clients ont également consulté
Slide 1 of 1
1 of 1
Alumina sheet, thickness 1.0 mm, size 100 × 100 mm, purity .%
GF04983497
Alumina