G3140
Glyoxal solution
40% in H2O
Synonym(s):
Ethanedial, Oxalaldehyde
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
Linear Formula:
OHCCHO
CAS Number:
Molecular Weight:
58.04
Beilstein:
1732463
EC Number:
MDL number:
UNSPSC Code:
12352103
Recommended Products
concentration
40% in H2O
density
1.27 g/mL at 25 °C (lit.)
SMILES string
[H]C(=O)C([H])=O
InChI
1S/C2H2O2/c3-1-2-4/h1-2H
InChI key
LEQAOMBKQFMDFZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
This form of glyoxal is composed of 3 moles of glyoxal and 2 moles of water in a relatively stable configuration.
Caution
A precipitate may form on storage which can be redissolved by warming to 50-60°C.
May precipitate on storage; redissolve at 50-60 °C.
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service